Learn More
enQuireBio™ Recombinant Human LRP4 Protein
A DNA sequence encoding the Homo sapiens (Human) LRP4, was expressed in the hosts and tags indicated.
Brand:  enQuireBio™ QP6320-ye-100ug
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 16072450
  • 10410.00 SEK / 100µg
Estimated Shipment: 17-06-2024
to see stock.
Specifications
NP_002325.2
LRP4 Protein
Research Use Only
LRP4
Recombinant Protein
Yeast
YRHKKSKFTD PGMGNLTYSN PSYRTSTQEV KIEAIPKPAM YNQLCYKKEG GPDHNYTKEK IKIVEGICLL SGDDAEWDDL KQLRSSRGGL LRDHVCMKTD TVSIQASSGS LDDTETEQLL QEEQSECSSV HTAATPERRG SLPDTGWKHE RKLSSESQV
Greater than 95% as determined by SDS-PAGE.
4038
100 μg
Not determined.
Cytoplasmic Domain
Human
His
Tris-based buffer, 50% glycerol