missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Lysozyme-like protein 1
A DNA sequence encoding the Homo sapiens (Human) Lysozyme-like protein 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7748-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_115906.3 | |
Lysozyme-like protein 1 | |
Research Use Only | |
LYZL1 | |
Recombinant Protein | |
E. coli | |
MQDAPLSCLS PTKWSSVSSA DSTEKSASGA GTRNLPFQFC LRQALRMKAA GILTLIGCLV TGAESKIYTR CKLAKIFSRA GLDNYWGFSL GNWICMAYYE SGYNTTAQTV LDDGSIDYGI FQINSFAWCR RGKLKENNHC HVACSALITD DLTDAIICAR KIVKETQGMN YWQGWKKHCE GRDLSEWKKG CEVS | |
Greater than 90% as determined by SDS-PAGE. |
84569 | |
500 μg | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |