missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human M-CSF / CSF-1 Protein
A DNA sequence encoding the Homo sapiens (Human) M-CSF / CSF-1, was expressed in the hosts and tags indicated.
2005.00 SEK - 21365.00 SEK
Specifications
Accession Number | NP_000748.3 |
---|---|
Gene ID (Entrez) | 1435 |
Name | M-CSF / CSF-1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16047591
|
enQuireBio™
QP9081-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16087591
|
enQuireBio™
QP9081-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16037591
|
enQuireBio™
QP9081-YE-100UG |
100 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16067591
|
enQuireBio™
QP9081-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16077591
|
enQuireBio™
QP9081-YE-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16057591
|
enQuireBio™
QP9081-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_000748.3 | |
M-CSF / CSF-1 Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
1435 | |
Research Use Only | |
CSF1 | |
Recombinant Protein | |
Yeast | |
EEVSEYCSHM IGSGHLQSLQ RLIDSQMETS CQITFEFVDQ EQLKDPVCYL KKAFLLVQDI MEDTMRFRDN TPNAIAIVQL QELSLRLKSC FTKDYEEHDK ACVRTFYETP LQLLEKVKNV FNETKNLLDK DWNIFSKNCN NSFAECSSQD VVTKPDCNCL YPKAIPSSDP ASVSPHQPLA PSMAPVAGLT WEDSEGTEGS SLLPGEQPLH TVDPGSAKQR PPRSTCQSFE PPETPVVKDS TIGGSPQPRP SVGAFNPGME DILDSAMGTN WVPEEASGEA SEIPVPQGTE LSPSRPGGGS MQTEPARPSN FLSASSPLPA SAKGQQPADV TGTALPRVGP VRPTGQDWNH TPQKTDHPSA LLRDPPEPGS PRISSLRPQG LSNPSTLSAQ PQLSRSHSSG SVLPLGELEG RRSTRDRRSP AEPEGGPASE GAARPLPRFN SVPLTDTGHE RQSEGSFSPQ LQESVFHLLV PSVILVLLAV GGLLFYRWRR RSHQEPQRAD SPLEQPEGSP LTQDDRQVEL PV | |
Greater than 90% as determined by SDS-PAGE. |