missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human M-CSF / CSF-1 Protein
A DNA sequence encoding the Homo sapiens (Human) M-CSF / CSF-1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9081-ye-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_000748.3 | |
M-CSF / CSF-1 Protein | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
1435 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
CSF1 | |
Recombinant Protein | |
Yeast | |
EEVSEYCSHM IGSGHLQSLQ RLIDSQMETS CQITFEFVDQ EQLKDPVCYL KKAFLLVQDI MEDTMRFRDN TPNAIAIVQL QELSLRLKSC FTKDYEEHDK ACVRTFYETP LQLLEKVKNV FNETKNLLDK DWNIFSKNCN NSFAECSSQD VVTKPDCNCL YPKAIPSSDP ASVSPHQPLA PSMAPVAGLT WEDSEGTEGS SLLPGEQPLH TVDPGSAKQR PPRSTCQSFE PPETPVVKDS TIGGSPQPRP SVGAFNPGME DILDSAMGTN WVPEEASGEA SEIPVPQGTE LSPSRPGGGS MQTEPARPSN FLSASSPLPA SAKGQQPADV TGTALPRVGP VRPTGQDWNH TPQKTDHPSA LLRDPPEPGS PRISSLRPQG LSNPSTLSAQ PQLSRSHSSG SVLPLGELEG RRSTRDRRSP AEPEGGPASE GAARPLPRFN SVPLTDTGHE RQSEGSFSPQ LQESVFHLLV PSVILVLLAV GGLLFYRWRR RSHQEPQRAD SPLEQPEGSP LTQDDRQVEL PV |