missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human MAD2L1 / MAD2 Protein

A DNA sequence encoding the Homo sapiens (Human) MAD2L1 / MAD2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7568-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16049450

  • 5980.00 SEK / 100µg
Estimated Shipment: 05-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_002349.1
MAD2L1 / MAD2 Protein
Research Use Only
MAD2L1
Recombinant Protein
E. coli
ALQLSREQGI TLRGSAEIVA EFFSFGINSI LYQRGIYPSE TFTRVQKYGL TLLVTTDLEL IKYLNNVVEQ LKDWLYKCSV QKLVVVISNI ESGEVLERWQ FDIECDKTAK DDSAPREKSQ KAIQDEIRSV IRQITATVTF LPLLEVSCSF DLLIYTDKDL VVPEKWEESG PQFITNSEEV RLRSFTTTIH KVNSMVAYKI PVND
Greater than 90% as determined by SDS-PAGE.
4085
100 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers