missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MAGOH Protein
A DNA sequence encoding the Homo sapiens (Human) MAGOH, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6333-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_002361.1 | |
MAGOH Protein | |
Research Use Only | |
MAGOH | |
Recombinant Protein | |
E. coli | |
MESDFYLRYY VGHKGKFGHE FLEFEFRPDG KLRYANNSNY KNDVMIRKEA YVHKSVMEEL KRIIDDSEIT KEDDALWPPP DRVGRQELEI VIGDEHISFT TSKIGSLIDV NQSKDPEGLR VFYYLVQDLK CLVFSLIGLH FKIKPI | |
Greater than 80% as determined by SDS-PAGE. |
4116 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |