missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Mammaglobin-A Protein
A DNA sequence encoding the Homo sapiens (Human) Mammaglobin-A, was expressed in the hosts and tags indicated.
1580.00 SEK - 19050.00 SEK
Specifications
Accession Number | NP_002402.1 |
---|---|
Gene ID (Entrez) | 4250 |
Name | Mammaglobin-A Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16059600
|
enQuireBio™
QP7590-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16099600
|
enQuireBio™
QP7590-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16049600
|
enQuireBio™
QP7590-EC-100UG |
100 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16079600
|
enQuireBio™
QP7590-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16089600
|
enQuireBio™
QP7590-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16069600
|
enQuireBio™
QP7590-EC-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_002402.1 | |
Mammaglobin-A Protein | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
4250 | |
Research Use Only | |
SCGB2A2 | |
Recombinant Protein | |
E. coli | |
GSGCPLLENV ISKTINPQVS KTEYKELLQE FIDDNATTNA IDELKECFLN QTDETLSNVE VFMQLIYDSS LCDLF | |
Greater than 80% as determined by SDS-PAGE. |