missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MART-1 / MLANA Protein
A DNA sequence encoding the Homo sapiens (Human) MART-1 / MLANA, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7586-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_005502.1 | |
MART-1 / MLANA Protein | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His-B2M | |
Tris-based buffer, 50% glycerol |
2315 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
MLANA | |
Recombinant Protein | |
E. coli | |
MPREDAHFIY GYPKKGHGHS YTTAEEAAGI GILTVILGVL LLIGCWYCRR RNGYRALMDK SLHVGTQCAL TRRCPQEGFD HRDSKVSLQE KNCEPVVPNA PPAYEKLSAE QSPPPYSP |