missing translation for 'onlineSavingsMsg'
Läs mer

R&D Systems™ Recombinant Human Mesothelin His Alexa Fluor™ 647 Protein

Produktkod. 18757896
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18757896 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 18757896 Leverantör R&D Systems™ Leverantörsnummer AFR3265020
 Begränsad artikel
Beräknad leverans: 02-12-2026
Lägg i varukorgen
Lägg i varukorgen

missing translation for 'validMainframeMsg'

Denna artikel kan inte returneras. Se returpolicy

C-Terminal (aa 296-580)

Measured by flow cytometry for its ability to bind anti-human Mesothelin Monoclonal Antibody conjugated beads.The concentration of Recombinant Human Mesothelin C-Terminal (aa 296-580) His-tag Alexa Fluor™ 647 (Catalog # AFR3265) that produces 50% of the binding response is 1.00-20.0 ng/mL.
TRUSTED_SUSTAINABILITY

Specifikationer

Tillträdesnummer AAH09272.1
För användning med (applikation) Bioactivity
Formulering Supplied as a 0.2 μm filtered solution in PBS with BSA as a carrier protein.
Gen-ID (Entrez) 10232
Molekylvikt (g/mol) M.W.-observed: 37-45 kDa, reducing conditions., M.W.-theroretical: 33 kDa
Kvantitet 20 μg
Källa Mouse myeloma cell line, NS0-derived human Mesothelin protein Glu296-Gly580, with a C-terminal 6-His tag
Förvaringskrav Protect from light. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after opening. 3 months, -20°C to -70°C under sterile conditions after opening.
Endotoxinkoncentration <1.0 EU / 1 μg of the protein by the LAL method.
Gene Alias CAK1, CAK1 antigen, megakaryocyte potentiating factor, mesothelin, MPF, MPFSMRP, MSLN, Pre-pro-megakaryocyte-potentiating factor, SMR, soluble MPF mesothelin related protein
Konjugera Alexa Fluor 647
Produktlinje R&D Systems
Renhet eller kvalitetsklass >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein Mesothelin
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.