missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Metalloreductase STEAP1 Protein
A DNA sequence encoding the Homo sapiens (Human) Metalloreductase STEAP1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8762-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_036581.1 | |
Metalloreductase STEAP1 Protein | |
Research Use Only | |
STEAP1 | |
Recombinant Protein | |
E. coli | |
SRKDITNQEE LWKMKPRRNL EEDDYLHKDT GETSMLKRPV LLHLHQTAHA DEFDCPSELQ HTQELFP | |
Greater than 90% as determined by SDS-PAGE. |
26872 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |