missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Metalloreductase STEAP1 Protein

A DNA sequence encoding the Homo sapiens (Human) Metalloreductase STEAP1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8762-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16006351

  • 22640.00 SEK / 1mg
Estimated Shipment: 12-07-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_036581.1
Metalloreductase STEAP1 Protein
1 mg
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol
26872
Greater than 90% as determined by SDS-PAGE.
Research Use Only
STEAP1
Recombinant Protein
E. coli
SRKDITNQEE LWKMKPRRNL EEDDYLHKDT GETSMLKRPV LLHLHQTAHA DEFDCPSELQ HTQELFP
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers