missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Metallothionein-1A Protein
A DNA sequence encoding the Homo sapiens (Human) Metallothionein-1A, was expressed in the hosts and tags indicated.
1580.00 SEK - 18390.00 SEK
Specifications
Accession Number | NP_005937.2 |
---|---|
Gene ID (Entrez) | 4489 |
Name | Metallothionein-1A Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16027071
|
enQuireBio™
QP8898-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16067071
|
enQuireBio™
QP8898-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16017071
|
enQuireBio™
QP8898-EC-100UG |
100 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16047071
|
enQuireBio™
QP8898-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16057071
|
enQuireBio™
QP8898-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16037071
|
enQuireBio™
QP8898-EC-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_005937.2 | |
Metallothionein-1A Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
4489 | |
Research Use Only | |
MT1A | |
Recombinant Protein | |
E. coli | |
MDPNCSCATG GSCTCTGSCK CKECKCTSCK KSCCSCCPMS CAKCAQGCIC KGASEKCSC | |
Greater than 90% as determined by SDS-PAGE. |