missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Metallothionein-3 Protein

A DNA sequence encoding the Homo sapiens (Human) Metallothionein-3, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6390-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16002970

  • 4960.00 SEK / 100µg
Estimated Shipment: 12-07-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_005945.1
Metallothionein-3 Protein
100 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
4504
Greater than 90% as determined by SDS-PAGE.
Research Use Only
MT3
Recombinant Protein
E. coli
MDPETCPCPS GGSCTCADSC KCEGCKCTSC KKSCCSCCPA ECEKCAKDCV CKGGEAAEAE AEKCSCCQ
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers