missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Midkine / MDK Protein
A DNA sequence encoding the Homo sapiens (Human) Midkine / MDK Protein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8694-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001012333.1 | |
Midkine / MDK Protein | |
Research Use Only | |
MDK | |
Recombinant Protein | |
E. coli | |
VAKKKDKVKK GGPGSECAEW AWGPCTPSSK DCGVGFREGT CGAQTQRIRC RVPCNWKKEF GADCKYKFEN WGACDGGTGT KVRQGTLKKA RYNAQCQETI RVTKPCTPKT KAKAKAKKGK GKD | |
Greater than 90% as determined by SDS-PAGE. |
4192 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |