missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Minpp1 Protein
A DNA sequence encoding the Homo sapiens (Human) Minpp1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8249-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001171588.1 | |
Minpp1 Protein | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
9562 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
Minpp1 | |
Recombinant Protein | |
E. coli | |
SLLEPRDPVA SSLSPYFGTK TRYEDVNPVL LSGPEAPWRD PELLEGTCTP VQLVALIRHG TRYPTVKQIR KLRQLHGLLQ ARGSRDGGAS STGSRDLGAA LADWPLWYAD WMDGQLVEKG RQDMRQLALR LASLFPALFS RENYGRLRLI TSSKHRCMDS SAAFLQGLWQ HYHPGLPPPD VADMEFGPPT VNDKLMRFFD HCEKFLTEVE KNATALYHVE AFKTGPEMQN ILKKVAATLQ VPVNDLNADL IQVAFFTCSF DLAIKGVKSP WCDVFDIDDA KVLEYLNDLK QYWKRGYGYT INSRSSCTLF QDIFQHLDKA VEQKQRSQPI SSPVILQFGH AETLLPLLSL MGYFKDKEPL TAYNYKKQMH RKFRSGLIVP YASNLIFVLY HCENAKTPKE QFRVQMLLNE KVLPLAYSQE TVSFYEDLKN HYKDILQSCQ TSEECELARA NSTSDEL |