missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Mitochondrial fission process protein 1

A DNA sequence encoding the Homo sapiens (Human) Mitochondrial fission process protein 1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8083-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16062561

  • 38570.00 SEK / 1mg
Estimated Shipment: 31-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001003704.1
Mitochondrial fission process protein 1
Research Use Only
MTFP1
Recombinant Protein
E. coli
MSEPQPRGAE RDLYRDTWVR YLGYANEVGE AFRSLVPAAV VWLSYGVASS YVLADAIDKG KKAGEVPSPE AGRSARVTVA VVDTFVWQAL ASVAIPGFTI NRVCAASLYV LGTATRWPLA VRKWTTTALG LLTIPIIIHP IDRSVDFLLD SSLRKLYPTV GKPSSS
Greater than 80% as determined by SDS-PAGE.
51537
1 mg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers