missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Mitochondrial import receptor subunit TOM34 Protein
A DNA sequence encoding the Homo sapiens (Human) Mitochondrial import receptor subunit TOM34, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7602-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_006800.2 | |
Mitochondrial import receptor subunit TOM34 Protein | |
Research Use Only | |
TOMM34 | |
Recombinant Protein | |
E. coli | |
MAPKFPDSVE ELRAAGNESF RNGQYAEASA LYGRALRVLQ AQGSSDPEEE SVLYSNRAAC HLKDGNCRDC IKDCTSALAL VPFSIKPLLR RASAYEALEK YPMAYVDYKT VLQIDDNVTS AVEGINRMTR ALMDSLGPEW RLKLPSIPLV PVSAQKRWNS LPSENHKEMA KSKSKETTAT KNRVPSAGDV EKARVLKEEG NELVKKGNHK KAIEKYSESL LCSNLESATY SNRALCYLVL KQYTEAVKDC TEALKLDGKN VKAFYRRAQA HKALKDYKSS FADISNLLQI EPRNGPAQKL RQEVKQNLH | |
Greater than 80% as determined by SDS-PAGE. |
10953 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |