missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Mitogen-activated protein kinase 6 Protein
A DNA sequence encoding the Homo sapiens (Human) Mitogen-activated protein kinase 6, was expressed in the hosts and tags indicated.
1640.00 SEK - 26990.00 SEK
Specifications
Accession Number | NP_002739.1 |
---|---|
Gene ID (Entrez) | 5597 |
Name | Mitogen-activated protein kinase 6 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16042600
|
enQuireBio™
QP6341-EC-100UG |
100 μg |
5980.00 SEK
100µg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16062600
|
enQuireBio™
QP6341-EC-1MG |
1 mg |
26990.00 SEK
1mg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16052600
|
enQuireBio™
QP6341-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16072600
|
enQuireBio™
QP6341-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16082600
|
enQuireBio™
QP6341-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16092600
|
enQuireBio™
QP6341-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_002739.1 | |
Mitogen-activated protein kinase 6 Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
5597 | |
Research Use Only | |
MAPK6 | |
Recombinant Protein | |
E. coli | |
MAEKFESLMN IHGFDLGSRY MDLKPLGCGG NGLVFSAVDN DCDKRVAIKK IVLTDPQSVK HALREIKIIR RLDHDNIVKV FEILGPSGSQ LTDDVGSLTE LNSVYIVQEY METDLANVLE QGPLLEEHAR LFMYQLLRGL KYIHSANVLH RDLKPANLFI NTEDLVLKIG DFGLARIMDP HYSHKGHLSE GLVTKWYRSP RLLLSPNNYT KAIDMWAAGC IFAEMLTGKT LFAGAHELEQ MQLILESIPV VHEEDRQELL SVIPVYIRND MTEPHKPLTQ LLPGISREAL DFLEQILTFS PMDRLTAEEA LSHPYMSIYS FPMDEPISSH PFHIEDEVDD ILLMDETHSH IYNWERYHDC QFSEHDWPVH NNFDIDEVQL DPRALSDVTD EEEVQVDPRK YLDGDREKYL EDPAFDTNYS TEPCWQYSDH HENKYCDLEC SHTCNYKTRS SSYLDNLVWR ESEVNHYYEP KLIIDLSNWK EQSKEKSDKK GKSKCERNGL VKAQIALEEA SQQLAGKERE KNQGFDFDSF IAGTIQLSSQ HEPTDVVDKL NDLNSSVSQL ELKSLISKSV SQEKQEKGMA NLAQLEALYQ SSWDSQFVSG GEDCFFINQF CEVRKDEQVE KENTYTSYLD KFFSRKEDTE MLETEPVEDG KLGERGHEEG FLNNSGEFLF NKQLESIGIP QFHSPVGSPL KSIQATLTPS AMKSSPQIPH QTYSSILKHL N | |
Greater than 90% as determined by SDS-PAGE. |