missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MMP28 Protein
A cDNA sequence encoding the MMP28 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12710-20ug
Additional Details : Weight : 0.01000kg
Specifications
MMP28 Protein | |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The MMP28 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol. |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSFAKQGNK WYKQHLSYRL VNWPEHLPEP AVRGAVRAAF QLWSNVSALE FWEAPATGPA DIRLTFFQGD HNDGLGNAFD GPGGALAHAF LPRRGEAHFD QDERWSLSRR RGRNLFVVLA HEIGHTLGLT HSPAPRALMA PYYKRLGRDA LLSWDDVLAV QSLYGKPLGG SVAVQLPGKL FTDFETWDSY SPQGRRPETQ GPKYCHSSFD AITVDRQQQL YIFKGSHFWE VAADGNVSEP RPLQERWVGL PPNIEAAAVS LNDGDFYFFK GGRCWRFRGP KPVWGLPQLC RAGGLPRHPD AALFFPPLRR LILFKGARYY VLARGGLQVE PYYPRSLQDW GGIPEEVSGA LPRPDGSIIF FRDDRYWRLD QAKLQATTSG RWATELPWMG CWHANSGSAL F |