missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MORF4 family-associated protein 1-like 1 Protein
A DNA sequence encoding the Homo sapiens (Human) MORF4 family-associated protein 1-like 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6379-ye-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_982287.1 | |
MORF4 family-associated protein 1-like 1 Protein | |
Research Use Only | |
MRFAP1L1 | |
Recombinant Protein | |
Yeast | |
MRPLDIDEVE APEEVEVLEP EEDFEQFLLP VINEMREDIA SLIREHGRAY LRTRSKLWEM DNMLIQIKTQ VEASEESALN HVQHPSGEAD ERVSELCEKA EEKAKEIAKM AEMLVELVWR IERSESS | |
Greater than 90% as determined by SDS-PAGE. |
114932 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |