Läs mer
enQuireBio™ Recombinant Human mRNA-decapping enzyme 1A Protein
Produktkod. 16053241
Kvantitet:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Förpackningsstorlek:
100µg
10µg
1mg
200µg
500µg
50µg
Produktkod. 16053241
Brand: enQuireBio™ QP8201ec200ug
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
A DNA sequence encoding the Homo sapiens (Human) mRNA-decapping enzyme 1A, was expressed in the hosts and tags indicated.
Specifikationer
Tillträdesnummer NP_001277133.1
Gen-ID (Entrez) 55802
Namn mRNA-decapping enzyme 1A Protein
Kvantitet 200 μg
Regulatorisk status Research Use Only
Endotoxinkoncentration Not determined.
Gensymbol DCP1A
Protein Längd Full Length
Produkttyp Recombinant Protein
Korsreaktivitet Human
Art E. coli
Protein Tag His
Sekvens MEALSRAGQE MSLAALKQHD PYITSIADLT GQVALYTFCP KANQWEKTDI EGTLFVYRRS ASPYHGFTIV NRLNMHNLVE PVNKDLEFQL HEPFLLYRNA SLSIYSIWFY DKNDCHRIAK LMADVVEEET RRSQQAARDK QSPSQANGCS DHRPIDILEM LSRAKDEYER NQMGDSNISS PGLQPSTQLS NLGSTETLEE MPSGSQDKSA PSGHKHLTVE ELFGTSLPKE QPAVVGLDSE EMERLPGDAS QKEPNSFLPF PFEQLGGAPQ SETLGVPSAA HHSVQPEITT PVLITPASIT QSNEKHAPTY TIPLSPVLSP TLPAEAPTAQ VPPSLPRNST MMQAVKTTPR QRSPLLNQPV PELSHASLIA NQSPFRAPLN VTNTAGTSLP SVDLLQKLRL TPQHDQIQTQ PLGKGAMVAS FSPAAGQLAT PESFIEPPSK TAAARVAASA SLSNMVLAPL QSMQQNQDPE VFVQPKVLSS AIQVAGAPLV TATTTAVSSV LLAPSVFQQT VTRSSDLERK ASSPSPLTIG TPESQRKPSI ILSKSQLQDT LIHLIKNDSS FLSTLHEVYL QVLTKNKDNH NL
Buffert Tris-based buffer, 50% glycerol
Renhet eller kvalitetsklass Greater than 90% as determined by SDS-PAGE.