missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Mucosal addressin cell adhesion molecule 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Mucosal addressin cell adhesion molecule 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7559-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_570116.2 | |
Mucosal addressin cell adhesion molecule 1 Protein | |
200 μg | |
Not determined. | |
Extracellular Domain | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
8174 | |
Greater than 80% as determined by SDS-PAGE. | |
Research Use Only | |
Madcam1 | |
Recombinant Protein | |
E. coli | |
QSLQVKPLQV EPPEPVVAVA LGASRQLTCR LACADRGASV QWRGLDTSLG AVQSDTGRSV LTVRNASLSA AGTRVCVGSC GGRTFQHTVQ LLVYAFPDQL TVSPAALVPG DPEVACTAHK VTPVDPNALS FSLLVGGQEL EGAQALGPEV QEEEEEPQGD EDVLFRVTER WRLPPLGTPV PPALYCQATM RLPGLELSHR QAIPVLHSPT SPEPPDTTSP ESPDTTSPES PDTTSQEPPD TTSPEPPDKT SPEPAPQQGS THTPRSPGST RTRRPEISQA GPTQGEVIPT GSSKPAGDQ |