missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Multidrug resistance protein 1
A DNA sequence encoding the Homo sapiens (Human) Multidrug resistance protein 1, was expressed in the hosts and tags indicated.
1640.00 SEK - 19050.00 SEK
Specifications
Accession Number | NP_000918.2 |
---|---|
Gene ID (Entrez) | 5243 |
Name | Multidrug resistance protein 1 |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16036161
|
enQuireBio™
QP8727-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16076161
|
enQuireBio™
QP8727-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16026161
|
enQuireBio™
QP8727-EC-100UG |
100 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16056161
|
enQuireBio™
QP8727-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16066161
|
enQuireBio™
QP8727-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16046161
|
enQuireBio™
QP8727-EC-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_000918.2 | |
Multidrug resistance protein 1 | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
5243 | |
Research Use Only | |
ABCB1 | |
Recombinant Protein | |
E. coli | |
LSSFTDKELL AYAKAGAVAE EVLAAIRTVI AFGGQKKELE RYNKNLEEAK RIGIKKAITA NI | |
Greater than 90% as determined by SDS-PAGE. |