missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Muscleblind-like protein 1
A DNA sequence encoding the Homo sapiens (Human) Muscleblind-like protein 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8222-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001300986.1 | |
Muscleblind-like protein 1 | |
Research Use Only | |
MBNL1 | |
Recombinant Protein | |
E. coli | |
MAVSVTPIRD TKWLTLEVCR EFQRGTCSRP DTECKFAHPS KSCQVENGRV IACFDSLKGR CSRENCKYLH PPPHLKTQLE INGRNNLIQQ KNMAMLAQQM QLANAMMPGA PLQPVPMFSV APSLATNASA AAFNPYLGPV SPSLVPAEIL PTAPMLVTGN PGVPVPAAAA AAAQKLMRTD RLEVCREYQR GNCNRGENDC RFAHPADSTM IDTNDNTVTV CMDYIKGRCS REKCKYFHPP AHLQAKIKAA QYQVNQAAAA QAAATAAAMG IPQAVLPPLP KRPALEKTNG ATAVFNTGIF QYQQALANMQ LQQHTAFLPP GSILCMTPAT SVVPMVHGAT PATVSAATTS ATSVPFAATA TANQIPIISA EHLTSHKYVT QM | |
Greater than 90% as determined by SDS-PAGE. |
4154 | |
1 mg | |
Not determined. | |
Full Length of Isoform 5 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |