missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Myo Protein
A cDNA sequence encoding the Myo was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12775-1mg
Additional Details : Weight : 0.01000kg
Specifications
4151 | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGLSDGEWQL VLNVWGKVEA DIPGHGQEVL IRLFKGHPET LEKFDKFKHL KSEDEMKASE DLKKHGATVL TALGGILKKK GHHEAEIKPL AQSHATKHKI PVKYLEFISE CIIQVLQSKH PGDFGADAQG AMNKALELFR KDMASNYKEL GFQG | |
Greater than 95.0% as determined by SDS-PAGE. |
Myo Protein | |
Research Use Only | |
MB | |
Human | |
Untagged | |
The sterile solution contains phosphate-buffered saline (pH 8.0) and 50mM phosphate-borate. |