missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Myoglobin His Protein
A cDNA sequence encoding the Myoglobin His was constructed and used to recombinantly synthesize the protein.
1545.00 SEK - 61725.00 SEK
Specifications
Gene ID (Entrez) | 4151 |
---|---|
Name | Myoglobin His Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | MB |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15961509
|
enQuireBio™
QP12777-5UG |
5 μg |
1545.00 SEK
5µg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15951509
|
enQuireBio™
QP12777-20UG |
20 μg |
2405.00 SEK
20µg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15941509
|
enQuireBio™
QP12777-1MG |
1 mg |
61725.00 SEK
1mg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
4151 | |
Research Use Only | |
MB | |
Human | |
His | |
The MB protein solution (1 mg/1ml) is formulated in 20mM Tris-HCl buffer (pH 8.0)1mM DTT, 100mM NaCl and 20% glycerol. |
Myoglobin His Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGLSDGEWQL VLNVWGKVEA DIPGHGQEVL IRLFKGHPET LEKFDKFKHL KSEDEMKASE DLKKHGATVL TALGGILKKK GHHEAEIKPL AQSHATKHKI PVKYLEFISE CIIQVLQSKH PGDFGADAQG AMNKALELFR KDMASNYKEL GFQG | |
Greater than 90% as determined by SDS-PAGE. |