missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Myomegalin Protein
A DNA sequence encoding the Homo sapiens (Human) Myomegalin, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7688-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001002810.1 | |
Myomegalin Protein | |
1 mg | |
Not determined. | |
Full Length of Isoform 8 | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
653513 | |
Greater than 80% as determined by SDS-PAGE. | |
Research Use Only | |
PDE4DIP | |
Recombinant Protein | |
E. coli | |
MKGTDSGSCC RRRCDFGCCC RASRRAHYTP YRSGDATRTP QSPRQTPSRE RRRPEPAGSW AAAAEEEEAA AAATPWMRDY FAEDDGEMVP RTSHTAAFLS DTKDRGPPVQ SQIWRSGEKV PFVQTYSLRA FEKPPQVQTQ ALRDFEKHLN DLKKENFSLK LRIYFLEERM QQKYEASRED IYKRNIELKV EVESLKRELQ DKKQHLDKTW ADVENLNSQN EAELRRQFEE RQQETEHVYE LLENKIQLLQ EESRLAKNEA ARMAALVEAE KECNLELSEK LKGVTKNWED VPGDQVKPDQ YTEALAQRDK |