missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human N-acetylmuramoyl-L-alanine amidase Protein
A DNA sequence encoding the Homo sapiens (Human) N-acetylmuramoyl-L-alanine amidase, was expressed in the hosts and tags indicated.
1580.00 SEK - 26990.00 SEK
Specifications
Accession Number | NP_443122.3 |
---|---|
Gene ID (Entrez) | 114770 |
Name | N-acetylmuramoyl-L-alanine amidase Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16051771
|
enQuireBio™
QP7947-EC-100UG |
100 μg |
5980.00 SEK
100µg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16071771
|
enQuireBio™
QP7947-EC-1MG |
1 mg |
26990.00 SEK
1mg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16061771
|
enQuireBio™
QP7947-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16081771
|
enQuireBio™
QP7947-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16091771
|
enQuireBio™
QP7947-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16001781
|
enQuireBio™
QP7947-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_443122.3 | |
N-acetylmuramoyl-L-alanine amidase Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
114770 | |
Research Use Only | |
PGLYRP2 | |
Recombinant Protein | |
E. coli | |
SLPLLMDSVI QALAELEQKV PAAKTRHTAS AWLMSAPNSG PHNRLYHFLL GAWSLNATEL DPCPLSPELL GLTKEVARHD VREGKEYGVV LAPDGSTVAV EPLLAGLEAG LQGRRVINLP LDSMAAPWET GDTFPDVVAI APDVRATSSP GLRDGSPDVT TADIGANTPD ATKGCPDVQA SLPDAKAKSP PTMVDSLLAV TLAGNLGLTF LRGSQTQSHP DLGTEGCWDQ LSAPRTFTLL DPKASLLTMA FLNGALDGVI LGDYLSRTPE PRPSLSHLLS QYYGAGVARD PGFRSNFRRQ NGAALTSASI LAQQVWGTLV LLQRLEPVHL QLQCMSQEQL AQVAANATKE FTEAFLGCPA IHPRCRWGAA PYRGRPKLLQ LPLGFLYVHH TYVPAPPCTD FTRCAANMRS MQRYHQDTQG WGDIGYSFVV GSDGYVYEGR GWHWVGAHTL GHNSRGFGVA IVGNYTAALP TEAALRTVRD TLPSCAVRAG LLRPDYALLG HRQLVRTDCP GDALFDLLRT WPHFTATVKP RPARSVSKRS RREPPPRTLP ATDLQ | |
Greater than 90% as determined by SDS-PAGE. |