missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human N-glycosylase / DNA lyase Protein
A DNA sequence encoding the Homo sapiens (Human) N-glycosylase/DNA lyase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6448-ye-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_002533.1 | |
N-glycosylase / DNA lyase Protein | |
Research Use Only | |
OGG1 | |
Recombinant Protein | |
Yeast | |
MPARALLPRR MGHRTLASTP ALWASIPCPR SELRLDLVLP SGQSFRWREQ SPAHWSGVLA DQVWTLTQTE EQLHCTVYRG DKSQASRPTP DELEAVRKYF QLDVTLAQLY HHWGSVDSHF QEVAQKFQGV RLLRQDPIEC LFSFICSSNN NIARITGMVE RLCQAFGPRL IQLDDVTYHG FPSLQALAGP EVEAHLRKLG LGYRARYVSA SARAILEEQG GLAWLQQLRE SSYEEAHKAL CILPGVGTKV ADCICLMALD KPQAVPVDVH MWHIAQRDYS WHPTTSQAKG PSPQTNKELG NFFRSLWGPY AGWAQAVLFS ADLRQSRHAQ EPPAKRRKGS KGPEG | |
Greater than 90% as determined by SDS-PAGE. |
4968 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |