missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human NAA38 Protein

A DNA sequence encoding the Homo sapiens (Human) NAA38, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8139-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16012861

  • 5980.00 SEK / 100µg
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001307854.1
NAA38 Protein
Research Use Only
NAA38
Recombinant Protein
E. coli
AVAVGVRAAP VLGLARALVL GLRGSQAARW RGWGTAAPGS LWALCECPAG CRELWFRGRA AWAARSERLL HPAQDSDGER EDSAAERARQ QLEALLNKTM RIRMTDGRTL VGCFLCTDRD CNVILGSAQE FLKPSDSFSA GEPRVLGLAM VPGHHIVSIE VQRESLTGPP YL
Greater than 90% as determined by SDS-PAGE.
84316
100 μg
Not determined.
Full Length of Isoform 2
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers