missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NAA38 Protein
A DNA sequence encoding the Homo sapiens (Human) NAA38, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8139-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001307854.1 | |
NAA38 Protein | |
Research Use Only | |
NAA38 | |
Recombinant Protein | |
E. coli | |
AVAVGVRAAP VLGLARALVL GLRGSQAARW RGWGTAAPGS LWALCECPAG CRELWFRGRA AWAARSERLL HPAQDSDGER EDSAAERARQ QLEALLNKTM RIRMTDGRTL VGCFLCTDRD CNVILGSAQE FLKPSDSFSA GEPRVLGLAM VPGHHIVSIE VQRESLTGPP YL | |
Greater than 90% as determined by SDS-PAGE. |
84316 | |
10 μg | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |