missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NAP-2 / PPBP / CXCL7 Protein
A DNA sequence encoding the Homo sapiens (Human) NAP-2 / PPBP / CXCL7, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8641-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_002695.1 | |
NAP-2 / PPBP / CXCL7 Protein | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
5473 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
PPBP | |
Recombinant Protein | |
E. coli | |
AELRCMCIKT TSGIHPKNIQ SLEVIGKGTH CNQVEVIATL KDGRKICLDP DAPRIKKIVQ KKLAGDE |