missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NDUFA12 Protein
A DNA sequence encoding the Homo sapiens (Human) NDUFA12, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8187-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001245267.1 | |
NDUFA12 Protein | |
Research Use Only | |
NDUFA12 | |
Recombinant Protein | |
E. coli | |
MELVQVLKRG LQQITGHGGL RGYLRVFFRT NDAKVGTLVG EDKYGNKYYE DNKQFFGRHR WVVYTTEMNG KNTFWDVDGS MVPPEWHRWL HSMTDDPPTT KPLTARKFIW TNHKFNVTGT PEQYVPYSTT RKKIQEWIPP STPYK | |
Greater than 80% as determined by SDS-PAGE. |
55967 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |