missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NDUFB5 Protein
A DNA sequence encoding the Homo sapiens (Human) NDUFB5, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6414-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001186886.1 | |
NDUFB5 Protein | |
Research Use Only | |
NDUFB5 | |
Recombinant Protein | |
E. coli | |
GQAELAEIPE GYVPEHWEYY KHPISRWIAR NFYDSPEKIY ERTMAVLQIE AEKAELRVKE LEVRKLMHVR GDGPWYYYET IDKELIDHSP KATPDN | |
Greater than 90% as determined by SDS-PAGE. |
4711 | |
1 mg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |