missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human NEIL1 Protein

A DNA sequence encoding the Homo sapiens (Human) NEIL1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7940-ec-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16081731

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001243481.1
NEIL1 Protein
Research Use Only
NEIL1
Recombinant Protein
E. coli
PEGPELHLAS QFVNEACRAL VFGGCVEKSS VSRNPEVPFE SSAYRISASA RGKELRLILS PLPGAQPQQE PLALVFRFGM SGSFQLVPRE ELPRHAHLRF YTAPPGPRLA LCFVDIRRFG RWDLGGKWQP GRGPCVLQEY QQFRENVLRN LADKAFDRPI CEALLDQRFF NGIGNYLRAE ILYRLKIPPF EKARSVLEAL QQHRPSPELT LSQKIRTKLQ NPDLLELCHS VPKEVVQLGG KGYGSESGEE DFAAFRAWLR CYGMPGMSSL QDRHGRTIWF QGDPGPLAPK GRKSRKKKSK ATQLSPEDRV EDALPPSKAP SRTRRAKRDL PKRTATQRPE GTSLQQDPEA PTVPKKGRRK GRQAASGHCR PRKVKADIPS LEPEGTSAS
Greater than 90% as determined by SDS-PAGE.
79661
50 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers