missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NKG2-C type II integral membrane Protein
A DNA sequence encoding the Homo sapiens (Human) NKG2-C type II integral membrane protein, was expressed in the hosts and tags indicated.
1580.00 SEK - 26990.00 SEK
Specifications
Accession Number | NP_002251.2 |
---|---|
Gene ID (Entrez) | 3822 |
Name | NKG2-C type II integral membrane Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16052080
|
enQuireBio™
QP6270-EC-100UG |
100 μg |
5980.00 SEK
100µg |
Estimated Shipment: 15-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16072080
|
enQuireBio™
QP6270-EC-1MG |
1 mg |
26990.00 SEK
1mg |
Estimated Shipment: 15-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16062080
|
enQuireBio™
QP6270-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16082080
|
enQuireBio™
QP6270-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16092080
|
enQuireBio™
QP6270-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16002090
|
enQuireBio™
QP6270-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_002251.2 | |
NKG2-C type II integral membrane Protein | |
Not determined. | |
Extracellular Domain | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
3822 | |
Research Use Only | |
KLRC2 | |
Recombinant Protein | |
E. coli | |
IPFLEQNNSS PNTRTQKARH CGHCPEEWIT YSNSCYYIGK ERRTWEESLL ACTSKNSSLL SIDNEEEMKF LASILPSSWI GVFRNSSHHP WVTINGLAFK HKIKDSDNAE LNCAVLQVNR LKSAQCGSSM IYHCKHKL | |
Greater than 90% as determined by SDS-PAGE. |