missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NKG2-C type II integral membrane Protein
A DNA sequence encoding the Homo sapiens (Human) NKG2-C type II integral membrane protein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6270-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_002251.2 | |
NKG2-C type II integral membrane Protein | |
Research Use Only | |
KLRC2 | |
Recombinant Protein | |
E. coli | |
IPFLEQNNSS PNTRTQKARH CGHCPEEWIT YSNSCYYIGK ERRTWEESLL ACTSKNSSLL SIDNEEEMKF LASILPSSWI GVFRNSSHHP WVTINGLAFK HKIKDSDNAE LNCAVLQVNR LKSAQCGSSM IYHCKHKL | |
Greater than 90% as determined by SDS-PAGE. |
3822 | |
10 μg | |
Not determined. | |
Extracellular Domain | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |