missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Beskrivning
This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.
Specifikationer
Specifikationer
| För användning med (applikation) | Western Blot, ELISA, Protein Array, Immunoaffinity Purification |
| Formulering | 50 mM Tris-HCl, 10 mM reduced Glutathione, pH 8.0 in the elution buffer. |
| Gen-ID (Entrez) | 388677 |
| Molekylvikt (g/mol) | MolecularWeight-theroretical: 51.7 kDa |
| Kvantitet | 25 μg |
| Förvaringskrav | Store at -80°C. Avoid freeze/thaw cycles. |
| Gene Alias | FLJ35032, FLJ55807, N2NNotch homolog 2 (Drosophila) N-terminal like, notch 2 N-terminal like, Notch homolog 2 N-terminal like protein, notch homolog 2 N-terminal-like protein |
| Gensymbol | NOTCH2NL |
| Forskningskategori | Growth and Development, Neuronal Cell Markers |
| Renhet eller kvalitetsklass | >80% by SDS-PAGE and Coomassie blue staining |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?