Läs mer
Novus Biologicals™ Recombinant Human NT5C3L His Protein
Produktkod. 18128009 Handla allt Bio Techne Produkter
Change view
Kvantitet:
0.1 mg
0.5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18128009 0.1 mg
18246490 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18128009 Leverantör Novus Biologicals™ Leverantörsnummer NBP2518960.1MG
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
Highly purified. Generating reliable and reproducible results.
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-300 of Human NT5C3L The Recombinant Human NT5C3L Protein is derived from E. coli. The Recombinant Human NT5C3L Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY
Specifikationer
Koncentration 1mg/mL
För användning med (applikation) SDS-PAGE
Formulering Phosphate buffered saline (pH7.4) containing 20% glycerol, 1mM DTT
Gen-ID (Entrez) 115024
Molekylvikt (g/mol) TMW: 36.8kDa
Kvantitet 0.1 mg
Förvaringskrav Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Gene Alias 5'-nucleotidase, cytosolic III-like
Vanligt namn NT5C3L
Konjugera Unconjugated
Rekombinant Recombinant
Renhet eller kvalitetsklass >85%, by SDS-PAGE
Visa mer Visa mindre
For research use only and is not approved for use in humans or in clinical diagnosis