missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NTF4 Protein
A DNA sequence encoding the Homo sapiens (Human) NTF4, was expressed in the hosts and tags indicated.
1640.00 SEK - 18390.00 SEK
Specifications
Accession Number | NP_006170.1 |
---|---|
Gene ID (Entrez) | 4909 |
Name | NTF4 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16035921
|
enQuireBio™
QP8678-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16075921
|
enQuireBio™
QP8678-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16025921
|
enQuireBio™
QP8678-EC-100UG |
100 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16055921
|
enQuireBio™
QP8678-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16065921
|
enQuireBio™
QP8678-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16045921
|
enQuireBio™
QP8678-EC-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_006170.1 | |
NTF4 Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
4909 | |
Research Use Only | |
NTF4 | |
Recombinant Protein | |
E. coli | |
VSETAPASRR GELAVCDAVS GWVTDRRTAV DLRGREVEVL GEVPAAGGSP LRQYFFETRC KADNAEEGGP GAGGGGCRGV DRRHWVSECK AKQSYVRALT ADAQGRVGWR WIRIDTACVC TLLSRTGRA | |
Greater than 90% as determined by SDS-PAGE. |