missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human NTF4 Protein

A DNA sequence encoding the Homo sapiens (Human) NTF4, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8678-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16065921

  • 11685.00 SEK / 500µg


Estimated Shipment
02-02-2021
Add to basket

Description

Description

Specifications

Specifications

NP_006170.1
NTF4 Protein
500 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
4909
Greater than 90% as determined by SDS-PAGE.
Research Use Only
NTF4
Recombinant Protein
E. coli
VSETAPASRR GELAVCDAVS GWVTDRRTAV DLRGREVEVL GEVPAAGGSP LRQYFFETRC KADNAEEGGP GAGGGGCRGV DRRHWVSECK AKQSYVRALT ADAQGRVGWR WIRIDTACVC TLLSRTGRA
Videos
SDS
Documents