missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Nuclear pore complex protein Nup153 Protein

A DNA sequence encoding the Homo sapiens (Human) Nuclear pore complex protein Nup153, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6439-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16023330

  • 22640.00 SEK / 1mg
Estimated Shipment: 01-11-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001265138.1
Nuclear pore complex protein Nup153 Protein
Research Use Only
NUP153
Recombinant Protein
E. coli
KAGSSWQCDT CLLQNKVTDN KCIACQAAKL SPRDTAKQTG IETPNKSGKT TLSASGTGFG DKFKPVIGTW DCDTCLVQNK PEAIKCVACE TPKPGTCVKR ALTLTVVSES AETMTASSSS CTVTTGTLGF GDKFKRPIGS WECSVCCVSN NAEDNKCVSC MSEKPGSSVP ASSSSTVPVS LPSGGSLGLE KFKKPEGSWD CELCLVQNKA DSTKCLACES AKPG
Greater than 90% as determined by SDS-PAGE.
9972
1 mg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers