missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Nuclear pore membrane glycoprotein 210 Protein
A DNA sequence encoding the Homo sapiens (Human) Nuclear pore membrane glycoprotein 210, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6440-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_079199.2 | |
Nuclear pore membrane glycoprotein 210 Protein | |
Research Use Only | |
NUP210 | |
Recombinant Protein | |
E. coli | |
AVGSVTVYYE VAGHLRTYKE VVVSVPQRIM ARHLHPIQTS FQEATASKVI VAVGDRSSNL RGECTPTQRE VIQALHPETL ISCQSQFKPA VFDFPSQDVF TVEPQFDTAL GQYFCSITMH RLTDKQRKHL SMKKTALVVS ASLSSSHFST EQVGAEVPFS PGLFADQAEI LLSNHYTSSE IRVFGAPEVL ENLEVKSGSP AVLAFAKEKS FGWPSFITYT VGVLDPAAGS QGPLSTTLTF SSPVTNQAIA IPVTVAFVVD RRGPGPYGAS LFQHFLDSYQ | |
Greater than 90% as determined by SDS-PAGE. |
23225 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |