Läs mer
enQuireBio™ Recombinant Human Obg-like ATPase 1 Protein
Produktkod. 16033410
Kvantitet:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Förpackningsstorlek:
100µg
10µg
1mg
200µg
500µg
50µg
Produktkod. 16033410
Brand: enQuireBio™ QP6451ec50ug
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
A DNA sequence encoding the Homo sapiens (Human) Obg-like ATPase 1, was expressed in the hosts and tags indicated.
Specifikationer
Tillträdesnummer NP_001011708.1
Gen-ID (Entrez) 29789
Namn Obg-like ATPase 1 Protein
Kvantitet 50 μg
Regulatorisk status Research Use Only
Endotoxinkoncentration Not determined.
Gensymbol OLA1
Protein Längd Full Length
Produkttyp Recombinant Protein
Korsreaktivitet Human
Art E. coli
Protein Tag His-SUMO
Sekvens MPPKKGGDGI KPPPIIGRFG TSLKIGIVGL PNVGKSTFFN VLTNSQASAE NFPFCTIDPN ESRVPVPDER FDFLCQYHKP ASKIPAFLNV VDIAGLVKGA HNGQGLGNAF LSHISACDGI FHLTRAFEDD DITHVEGSVD PIRDIEIIHE ELQLKDEEMI GPIIDKLEKV AVRGGDKKLK PEYDIMCKVK SWVIDQKKPV RFYHDWNDKE IEVLNKHLFL TSKPMVYLVN LSEKDYIRKK NKWLIKIKEW VDKYDPGALV IPFSGALELK LQELSAEERQ KYLEANMTQS ALPKIIKAGF AALQLEYFFT AGPDEVRAWT IRKGTKAPQA AGKIHTDFEK GFIMAEVMKY EDFKEEGSEN AVKAAGKYRQ QGRNYIVEDG DIIFFKFNTP QQPKKK
Buffert Tris-based buffer, 50% glycerol
Renhet eller kvalitetsklass Greater than 90% as determined by SDS-PAGE.