missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Orofacial cleft 1 candidate gene 1 Protein

A DNA sequence encoding the Homo sapiens (Human) Orofacial cleft 1 candidate gene 1 protein, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7807-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16060921

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

Orofacial cleft 1 candidate gene 1 Protein
Research Use Only
OFCC1
Recombinant Protein
E. coli
REKFQQKALK QTKQKKSKSA EFLMVKEDRE ATEGTGNPAF NMSSPDLSAC QTAEKKVIRH DMPDRTLAAH QQKFRLPASA EPKGNEYGRN YFDPLMDEEI NPRQCATEVS REDDDRIFYN RLTKLFDESR QGEPQDESGR EETLNSEAPG SSNKSHEIHK EASEATTAHL EEFQRSQKTI ILLGSSPLEQ EIRSTSLHCM EDEMSHPWIL LLKVTAVIRS RRYYREQRF
Greater than 90% as determined by SDS-PAGE.
200 μg
Not determined.
Partial (see sequence information for more details).
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers