Learn More
enQuireBio™ Recombinant Human OSGEP Protein
A DNA sequence encoding the Homo sapiens (Human) OSGEP, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8150-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_060277.1 | |
OSGEP Protein | |
Research Use Only | |
OSGEP | |
Recombinant Protein | |
E. coli | |
MPAVLGFEGS ANKIGVGVVR DGKVLANPRR TYVTPPGTGF LPGDTARHHR AVILDLLQEA LTESGLTSQD IDCIAYTKGP GMGAPLVSVA VVARTVAQLW NKPLVGVNHC IGHIEMGRLI TGATSPTVLY VSGGNTQVIA YSEHRYRIFG ETIDIAVGNC LDRFARVLKI SNDPSPGYNI EQMAKRGKKL VELPYTVKGM DVSFSGILSF IEDVAHRMLA TGECTPEDLC FSLQETVFAM LVEITERAMA HCGSQEALIV GGVGCNVRLQ EMMATMCQER GARLFATDER FCIDNGAMIA QAGWEMFRAG HRTPLSDSGV TQRYRTDEVE VTWRD | |
Greater than 80% as determined by SDS-PAGE. |
55644 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |