missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human p38 delta / MAPK13 Protein
A DNA sequence encoding the Homo sapiens (Human) p38 delta / MAPK13, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6339-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_002745.1 | |
p38 delta / MAPK13 Protein | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
5603 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
MAPK13 | |
Recombinant Protein | |
E. coli | |
MSLIRKKGFY KQDVNKTAWE LPKTYVSPTH VGSGAYGSVC SAIDKRSGEK VAIKKLSRPF QSEIFAKRAY RELLLLKHMQ HENVIGLLDV FTPASSLRNF YDFYLVMPFM QTDLQKIMGM EFSEEKIQYL VYQMLKGLKY IHSAGVVHRD LKPGNLAVNE DCELKILDFG LARHADAEMT GYVVTRWYRA PEVILSWMHY NQTVDIWSVG CIMAEMLTGK TLFKGKDYLD QLTQILKVTG VPGTEFVQKL NDKAAKSYIQ SLPQTPRKDF TQLFPRASPQ AADLLEKMLE LDVDKRLTAA QALTHPFFEP FRDPEEETEA QQPFDDSLEH EKLTVDEWKQ HIYKEIVNFS PIARKDSRRR SGMKL |