missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human P4HB / ERBA2L Protein
A DNA sequence encoding the Homo sapiens (Human) P4HB / ERBA2L, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6460-ye-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_000909.2 | |
P4HB / ERBA2L Protein | |
Research Use Only | |
P4hb | |
Recombinant Protein | |
Yeast | |
DAPEEEDHVL VLRKSNFAEA LAAHKYLLVE FYAPWCGHCK ALAPEYAKAA GKLKAEGSEI RLAKVDATEE SDLAQQYGVR GYPTIKFFRN GDTASPKEYT AGREADDIVN WLKKRTGPAA TTLPDGAAAE SLVESSEVAV IGFFKDVESD SAKQFLQAAE AIDDIPFGIT SNSDVFSKYQ LDKDGVVLFK KFDEGRNNFE GEVTKENLLD FIKHNQLPLV IEFTEQTAPK IFGGEIKTHI LLFLPKSVSD YDGKLSNFKT AAESFKGKIL FIFIDSDHTD NQRILEFFGL KKEECPAVRL ITLEEEMTKY KPESEELTAE RITEFCHRFL EGKIKPHLMS QELPEDWDKQ PVKVLVGKNF EDVAFDEKKN VFVEFYAPWC GHCKQLAPIW DKLGETYKDH ENIVIAKMDS TANEVEAVKV HSFPTLKFFP ASADRTVIDY NGERTLDGFK KFLESGGQDG AGDDDDLEDL EEAEEPDMEE DDDQKAVKDE L | |
Greater than 90% as determined by SDS-PAGE. |
5034 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |