missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PAH / PH Protein

A DNA sequence encoding the Homo sapiens (Human) PAH / PH, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6463-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16023480

  • 5980.00 SEK / 100µg
Estimated Shipment: 18-01-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_000268.1
PAH / PH Protein
Research Use Only
PAH
Recombinant Protein
E. coli
STAVLENPGL GRKLSDFGQE TSYIEDNCNQ NGAISLIFSL KEEVGALAKV LRLFEENDVN LTHIESRPSR LKKDEYEFFT HLDKRSLPAL TNIIKILRHD IGATVHELSR DKKKDTVPWF PRTIQELDRF ANQILSYGAE LDADHPGFKD PVYRARRKQF ADIAYNYRHG QPIPRVEYME EGKKTWGTVF KTLKSLYKTH ACYEYNHIFP LLEKYCGFHE DNIPQLEDVS QFLQTCTGFR LRPVAGLLSS RDFLGGLAFR VFHCTQYIRH GSKPMYTPEP DICHELLGHV PLFSDRSFAQ FSQEIGLASL GAPDEYIEKL ATIYWFTVEF GLCKQGDSIK AYGAGLLSSF GELQYCLSEK PKLLPLELEK TAIQNYTVTE FQPLYYVAES FNDAKEKVRN FAATIPRPFS VRYDPYTQRI EVLDNTQQLK ILADSINSEI GILCSALQKI K
Greater than 80% as determined by SDS-PAGE.
5053
100 μg
Not determined.
Full Length of Mature. Protein
Human
His-SUMO
Tris-based buffer, 50% glycerol
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers