missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Paralemmin-1 Protein
A DNA sequence encoding the Homo sapiens (Human) Paralemmin-1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6465-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001035224.1 | |
Paralemmin-1 Protein | |
Research Use Only | |
PALM | |
Recombinant Protein | |
E. coli | |
MEVLAAETTS QQERLQAIAE KRKRQAEIEN KRRQLEDERR QLQHLKSKAL RERWLLEGTP SSASEGDEDL RRQMQDDEQK TRLLEDSVSR LEKEIEVLER GDSAPATAKE NAAAPSPVRA PAPSPAKEER KTEVVMNSQQ TPVGTPKDKR VSNTPLRTVD GSPMMKAAMY SVEITVEKDK VTGETRVLSS TTLLPRQPLP LGIKVYEDET KVVHAVDGTA ENGIHPLSSS EVDELIHKAD EVTLSEAGST AGAAETRGAV EGAARTTPSR REITGVQAQP GEATSGPPGI QPGQEPPVTM IFMGYQNVED EAETKKVLGL QDTITAELVV IEDAAEPKEP APPNGSAAEP PTEAASREEN QAGPEATTSD PQDLDMKKHR CKCC | |
Greater than 90% as determined by SDS-PAGE. |
5064 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |